siRNA Details
Gene Symbol: Capn5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134461.1 | NP_604456.1 | 4 | 537 | ||
XM_006229729.2 | XP_006229791.2 | 4 | 774 | |||
XM_006229730.2 | XP_006229792.1 | 4 | 666 | |||
XM_006229731.2 | XP_006229793.1 | 4 | 910 | |||
GenBank mRNA | AF484958.1 | - | 4 | 537 | ||
FQ223532.1 | - | 4 | 631 |
