siRNA Details
Gene Symbol: LOC171573 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138537.2 | NP_612546.1 | 3 | 238 | ||
GenBank mRNA | AF198442.1 | - | 3 | 220 | ||
CB695439.1 | - | 3 | 259 | |||
FQ222501.1 | - | 3 | 258 | |||
FQ228610.1 | - | 3 | 258 |
