siRNA Details
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342494.7 | XP_342495.4 | 8, 9 | 1271 | ||
GenBank mRNA | EV775747.1 | - | 1, 2 | 75 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bub1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_342494.7 | XP_342495.4 | 8, 9 | 1271 | ||
GenBank mRNA | EV775747.1 | - | 1, 2 | 75 |