siRNA Details
Gene Symbol: Hyou1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034028.1 | NP_001029200.1 | 26 | 3017 | ||
NM_138867.2 | NP_620222.2 | 26 | 3169 | |||
XM_006242884.2 | XP_006242946.1 | 26 | 3114 | |||
XM_006242885.2 | XP_006242947.1 | 26 | 3050 | |||
XM_008766143.1 | XP_008764365.1 | 26 | 3096 | |||
GenBank mRNA | BC065310.1 | - | 26 | 3060 | 5621638 | |
U41853.1 | - | 26 | 3172 |
