siRNA Details
Gene Symbol: Mtnr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100641.1 | NP_001094111.1 | 1, 2 | 329 | ||
GenBank mRNA | AB377275.1 | - | 1, 2 | 329 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
