siRNA Details
Gene Symbol: Calm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012518.3 | NP_036650.1 | 4 | 408 | ||
GenBank mRNA | BC063187.1 | - | 503 | 6890866 | ||
FQ212716.1 | - | 474 | ||||
M16659.1 | - | 4 | 408 | |||
X13817.1 | - | 5 | 500 |
