siRNA Details
Gene Symbol: Gfap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017009.2 | NP_058705.2 | 1 | 237 | ||
XM_008768261.1 | XP_008766483.1 | 1 | 303 | |||
GenBank mRNA | BC088851.1 | - | 1 | 237 | 7315338 | |
EF094476.1 | - | 1 | 308 | |||
L27219.1 | - | 1 | 241 | |||
U03700.1 | - | 1 | 261 |
