siRNA Details
Gene Symbol: Nr3c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012576.2 | NP_036708.2 | 1 | 610 | ||
XM_008772065.1 | XP_008770287.1 | 1 | 611 | |||
XM_008772066.1 | XP_008770288.1 | 2 | 661 | |||
GenBank mRNA | AF455050.1 | - | 1 | 601 | ||
AY066016.2 | - | 1 | 605 | |||
M14053.1 | - | 1 | 668 | |||
Y00489.1 | - | 1 | 600 | |||
Y12264.1 | - | 1 | 622 |
