siRNA Details
Gene Symbol: Mbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025289.1 | NP_001020460.1 | 4 | 744 | ||
NM_001025291.1 | NP_001020462.1 | 1 | 184 | |||
NM_001025292.1 | NP_001020463.1 | 1 | 184 | |||
NM_001025293.1 | NP_001020464.1 | 1 | 184 | |||
NM_001025294.1 | NP_001020465.1 | 1 | 184 | |||
NM_017026.2 | NP_058722.1 | 1 | 184 | |||
XM_006254996.2 | XP_006255058.1 | 4 | 770 | |||
XM_006254997.2 | XP_006255059.1 | 4 | 846 | |||
XM_008772082.1 | XP_008770304.1 | 4 | 861 | |||
XM_008772083.1 | XP_008770305.1 | 3 | 401 | |||
GenBank mRNA | AF439750.1 | - | 1 | 107 | ||
AJ132895.1 | - | 1 | 107 | |||
AJ132896.1 | - | 1 | 107 | |||
AJ132897.1 | - | 1 | 107 | |||
AJ132898.1 | - | 1 | 107 | |||
BC084713.1 | - | 4 | 744 | 7303731 | ||
BC094522.1 | - | 1 | 140 | 7316167 | ||
CB586909.1 | - | 1 | 212 | |||
FQ214209.1 | - | 1 | 151 | |||
JX131663.1 | - | 1 | 184 | |||
JX131664.1 | - | 1 | 184 | |||
K00512.1 | - | 1 | 131 | |||
M25889.1 | - | 1 | 147 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001025245, NM_001025255, NM_001025259, NM_001025258, NM_001025251, and others.
Gene Ontology Categories:
