siRNA Details
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031517.2 | NP_113705.1 | 2 | 1366 | ||
XM_006236129.2 | XP_006236191.1 | 4 | 1782 | |||
GenBank mRNA | S69881.1 | - | 2 | 277 | ||
U65007.1 | - | 2 | 1366 | |||
X96786.1 | - | 2 | 1366 |
