siRNA Details
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031517.2 | NP_113705.1 | 16 | 3380 | ||
XM_006236129.2 | XP_006236191.1 | 18 | 3796 | |||
GenBank mRNA | U65007.1 | - | 16 | 3380 | ||
X96786.1 | - | 16 | 3380 |
