siRNA Details
Gene Symbol: Met | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031517.2 | NP_113705.1 | 16 | 3501 | ||
XM_006236129.2 | XP_006236191.1 | 18 | 3917 | |||
GenBank mRNA | AF352173.1 | - | 1 | 94 | ||
U65007.1 | - | 16 | 3501 | |||
X96786.1 | - | 16 | 3501 |
