siRNA Details
Gene Symbol: Cd200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031518.2 | NP_113706.2 | 5 | 844 | ||
XM_006248325.2 | XP_006248387.1 | 5 | 855 | |||
GenBank mRNA | BC089793.1 | - | 5 | 844 | 7380792 | |
X01785.1 | - | 5 | 801 |
