siRNA Details
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170600.1 | NP_001164071.1 | 17 | 2305 | ||
XM_006245842.2 | XP_006245904.1 | 19 | 2433 | |||
XM_006245844.2 | XP_006245906.1 | 19 | 2428 | |||
GenBank mRNA | AY953023.1 | - | 16 | 1746 | ||
BC166736.1 | - | 18 | 2268 |
