siRNA Details
Gene Symbol: Myh11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170600.1 | NP_001164071.1 | 38 | 5582 | ||
XM_006245842.2 | XP_006245904.1 | 40 | 5710 | |||
XM_006245844.2 | XP_006245906.1 | 40 | 5705 | |||
GenBank mRNA | X16261.1 | - | 4 | 457 | ||
X16262.1 | - | 9 | 1381 |
