siRNA Details
Gene Symbol: Myl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012606.2 | NP_036738.1 | 3, 4 | 372 | ||
XM_006243917.2 | XP_006243979.2 | 3, 4 | 473 | |||
GenBank mRNA | BC081832.1 | - | 3, 4 | 372 | 7192037 | |
FQ216106.1 | - | 3, 4 | 384 | |||
FQ216251.1 | - | 3, 4 | 384 | |||
FQ216255.1 | - | 3, 4 | 404 | |||
FQ217222.1 | - | 3, 4 | 387 | |||
FQ223547.1 | - | 3, 4 | 384 | |||
X14812.1 | - | 3, 4 | 365 |
