siRNA Details
Gene Symbol: Egfl7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139104.1 | NP_620804.1 | 6 | 814 | ||
XM_006233668.2 | XP_006233730.1 | 6 | 917 | |||
XM_006233669.2 | XP_006233731.1 | 5 | 471 | |||
GenBank mRNA | AF223678.1 | - | 6 | 814 | ||
BC078725.1 | - | 6 | 758 | 7127696 |
