siRNA Details
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012612.2 | NP_036744.1 | 2 | 504 | ||
GenBank mRNA | AI602287.1 | - | 2 | 324 | ||
BC158590.1 | - | 2 | 494 | 9036379 | ||
M15868.1 | - | 1 | 318 | |||
M27498.1 | - | 2 | 492 | |||
X00665.1 | - | 2 | 496 | |||
X01118.1 | - | 2 | 485 |
