siRNA Details
Gene Symbol: Odc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239906.2 | 8 | 1090 | |||
XM_006239907.2 | XP_006239969.1 | 8 | 1080 | |||
XM_006239908.2 | XP_006239970.1 | 7 | 996 | |||
XM_008764573.1 | 8 | 1063 | ||||
GenBank mRNA | BC078882.1 | - | 6 | 768 | 7115603 | |
J04791.1 | - | 8 | 884 | |||
M16982.1 | - | 8 | 1148 |
