siRNA Details
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136241.1 | NP_001129713.1 | 1110 | |||
GenBank mRNA | BC078722.1 | - | 1 | 106 | ||
BC091122.1 | - | 252 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ngef | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136241.1 | NP_001129713.1 | 1110 | |||
GenBank mRNA | BC078722.1 | - | 1 | 106 | ||
BC091122.1 | - | 252 |