siRNA Details
Gene Symbol: Macrod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139337.1 | NP_647553.1 | 5, 6 | 451 | ||
XM_006230677.2 | XP_006230739.1 | 5, 6 | 785 | |||
GenBank mRNA | AF404762.1 | - | 5, 6 | 451 | ||
FQ216840.1 | - | 5, 6 | 618 |
