siRNA Details
Gene Symbol: Tax1bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004199.1 | NP_001004199.1 | 2 | 185 | ||
XM_006236480.2 | XP_006236542.1 | 2 | 228 | |||
XM_006236481.2 | XP_006236543.1 | 2 | 671 | |||
XM_006236482.2 | XP_006236544.1 | 2 | 228 | |||
XM_006236483.2 | XP_006236545.1 | 2 | 228 | |||
GenBank mRNA | AY318959.1 | - | 101 | |||
BC081949.1 | - | 2 | 185 | 7122283 |
