siRNA Details
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144747.3 | NP_653348.3 | 5 | 1190 | ||
GenBank mRNA | AB075229.1 | - | 4 | 1099 | ||
BC093599.1 | - | 5 | 1190 | 7315729 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc45a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144747.3 | NP_653348.3 | 5 | 1190 | ||
GenBank mRNA | AB075229.1 | - | 4 | 1099 | ||
BC093599.1 | - | 5 | 1190 | 7315729 |