siRNA Details
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113570.1 | NP_001107042.1 | 3 | 470 | ||
GenBank mRNA | AF391798.1 | - | 3 | 231 | ||
CB616240.1 | - | 3 | 478 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113570.1 | NP_001107042.1 | 3 | 470 | ||
GenBank mRNA | AF391798.1 | - | 3 | 231 | ||
CB616240.1 | - | 3 | 478 |