siRNA Details
Gene Symbol: Gtf3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113570.1 | NP_001107042.1 | 7 | 910 | ||
GenBank mRNA | AF391798.1 | - | 7 | 671 | ||
CB578423.1 | - | 3 | 453 | |||
CK842762.1 | - | 3 | 435 |
