siRNA Details
Gene Symbol: Pgam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053290.1 | NP_445742.1 | 2 | 283 | ||
GenBank mRNA | BC065582.1 | - | 2 | 338 | 5623677 | |
M76591.1 | - | 2 | 283 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pgam1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053290.1 | NP_445742.1 | 2 | 283 | ||
GenBank mRNA | BC065582.1 | - | 2 | 338 | 5623677 | |
M76591.1 | - | 2 | 283 |