siRNA Details
Gene Symbol: Allc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013037.1 | NP_001013055.1 | 7 | 702 | ||
XM_006239944.2 | XP_006240006.1 | 7 | 776 | |||
GenBank mRNA | AF480062.1 | - | 4 | 356 | ||
BC078971.1 | - | 7 | 702 | 7111813 |
