siRNA Details
Gene Symbol: Mafk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145673.2 | NP_663706.2 | 3 | 334 | ||
XM_006248915.2 | XP_006248977.1 | 3 | 3575 | |||
XM_006248917.2 | XP_006248979.1 | 3 | 208 | |||
GenBank mRNA | AF461686.1 | - | 3 | 249 | ||
BC070906.1 | - | 3 | 334 | 7103075 |
