siRNA Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105713.1 | NP_001099183.1 | 16 | 2165 | ||
XM_008768310.1 | XP_008766532.1 | 17 | 2525 | |||
GenBank mRNA | BC169007.1 | - | 17 | 2222 | 7387040 | |
X07286.1 | - | 17 | 2224 |
