siRNA Details
Gene Symbol: Prkca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105713.1 | NP_001099183.1 | 6, 7 | 942 | ||
XM_008768310.1 | XP_008766532.1 | 6, 7 | 1179 | |||
GenBank mRNA | BC169007.1 | - | 6, 7 | 891 | 7387040 | |
X07286.1 | - | 6, 7 | 893 |

More Information
Other Species Targeted

Gene Ontology Categories:
