siRNA Details
Gene Symbol: Kpnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017063.1 | NP_058759.1 | 12 | 1593 | ||
GenBank mRNA | L38644.1 | - | 12 | 1593 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Kpnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017063.1 | NP_058759.1 | 12 | 1593 | ||
GenBank mRNA | L38644.1 | - | 12 | 1593 |