siRNA Details
Gene Symbol: Fgfr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109904.1 | NP_001103374.1 | 9 | 1373 | ||
XM_006253606.2 | XP_006253668.1 | 9 | 1390 | |||
XM_008771485.1 | XP_008769707.1 | 9 | 1384 | |||
GenBank mRNA | BC100260.1 | - | 8 | 1465 | 7100540 | |
M91599.1 | - | 6 | 839 |
