siRNA Details
Gene Symbol: Slc2a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012751.1 | NP_036883.1 | 1 | 159 | ||
XM_006246596.2 | XP_006246658.1 | 2 | 132 | |||
GenBank mRNA | BC085757.1 | - | 1 | 204 | 7192278 | |
D28561.1 | - | 1 | 159 | |||
FQ216450.1 | - | 2 | 98 | |||
FQ232657.1 | - | 1 | 218 | |||
FQ233419.1 | - | 1 | 194 | |||
J04524.1 | - | 1 | 141 | |||
KJ696745.1 | - | 1 | 44 | |||
M25482.1 | - | 1 | 124 | |||
X14771.1 | - | 1 | 172 |
