siRNA Details
Gene Symbol: Gata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033442.1 | NP_254277.1 | 3 | 577 | ||
GenBank mRNA | AB102789.1 | - | 1 | 134 | ||
AB102790.1 | - | 1 | 134 | |||
AF345897.1 | - | 1 | 134 | |||
AY032734.1 | - | 3 | 577 | |||
BC061745.1 | - | 2 | 384 | 5599740 | ||
FQ212304.1 | - | 458 |
