siRNA Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017093.1 | NP_058789.1 | 11 | 1427 | ||
XM_008759108.1 | XP_008757330.1 | 12 | 3826 | |||
XM_008759109.1 | XP_008757331.1 | 11 | 1414 | |||
XM_008759110.1 | XP_008757332.1 | 11 | 3691 | |||
XM_008759111.1 | XP_008757333.1 | 11 | 1537 | |||
GenBank mRNA | D30041.1 | - | 11 | 1427 | ||
DQ198085.1 | - | 10 | 1146 |
