siRNA Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017093.1 | NP_058789.1 | 13, 14 | 1648 | ||
XM_008759108.1 | XP_008757330.1 | 14, 15 | 4047 | |||
XM_008759109.1 | XP_008757331.1 | 13, 14 | 1635 | |||
XM_008759110.1 | XP_008757332.1 | 13, 14 | 3912 | |||
XM_008759111.1 | XP_008757333.1 | 13, 14 | 1758 | |||
GenBank mRNA | D30041.1 | - | 13, 14 | 1648 | ||
DQ198085.1 | - | 12, 13 | 1367 |
