siRNA Details
Gene Symbol: Mapk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270556.1 | NP_001257485.1 | 6, 7 | 926 | ||
NM_012806.2 | NP_036938.2 | 6, 7 | 953 | |||
XM_008769992.1 | XP_008768214.1 | 5, 6 | 3745 | |||
GenBank mRNA | DQ377224.1 | - | 3, 4 | 300 | ||
FQ211777.1 | - | 6, 7 | 925 | |||
L27128.1 | - | 6, 7 | 663 |
