siRNA Details
Gene Symbol: Cox6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012814.1 | NP_036946.1 | 2 | 235 | ||
GenBank mRNA | CB716915.1 | - | 2 | 235 | ||
FQ211611.1 | - | 2 | 222 | |||
FQ213920.1 | - | 2 | 222 | |||
FQ221129.1 | - | 2 | 227 | |||
FQ221354.1 | - | 2 | 222 | |||
FQ221968.1 | - | 2 | 224 | |||
FQ224366.1 | - | 2 | 222 | |||
FQ225833.1 | - | 2 | 222 | |||
X12553.1 | - | 2 | 136 |
