siRNA Details
Gene Symbol: Tsga10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030022.1 | NP_001025193.1 | 17 | 2166 | ||
GenBank mRNA | AF092091.1 | - | 17 | 2166 | ||
BC099189.1 | - | 2 | 464 | |||
BC168202.1 | - | 16 | 2094 | 7115410 |
