siRNA Details
Gene Symbol: Cck | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012829.2 | NP_036961.1 | 3 | 433 | ||
XM_006244070.2 | XP_006244132.1 | 4 | 697 | |||
GenBank mRNA | M25942.1 | - | 3 | 356 | ||
X01032.1 | - | 3 | 433 |
