siRNA Details
Gene Symbol: Mat1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012860.2 | NP_036992.2 | 9 | 1285 | ||
GenBank mRNA | BC089770.1 | - | 9 | 1285 | 7368255 | |
X15734.1 | - | 9 | 1195 | |||
X60822.1 | - | 9 | 1371 |
