siRNA Details
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012911.1 | NP_037043.1 | 12 | 1137 | ||
GenBank mRNA | BC087578.1 | - | 12 | 1009 | 7311618 | |
M91590.1 | - | 12 | 1137 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Arrb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012911.1 | NP_037043.1 | 12 | 1137 | ||
GenBank mRNA | BC087578.1 | - | 12 | 1009 | 7311618 | |
M91590.1 | - | 12 | 1137 |