siRNA Details
Gene Symbol: Eef2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012947.2 | NP_037079.1 | 6 | 692 | ||
XM_006230156.2 | XP_006230218.1 | 7 | 1016 | |||
XM_006230157.2 | XP_006230219.1 | 7 | 1413 | |||
XM_006230158.2 | XP_006230220.1 | 7 | 2093 | |||
GenBank mRNA | BC061825.1 | - | 7 | 947 | 5599075 | |
U93849.1 | - | 6 | 692 | |||
X96426.1 | - | 982 |
