siRNA Details
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017098.1 | NP_058794.1 | 2, 3 | 286 | ||
XM_008767638.1 | XP_008765860.1 | 1, 2 | 358 | |||
GenBank mRNA | L22788.1 | - | 2, 3 | 286 | ||
S52878.1 | - | 2, 3 | 265 |
