siRNA Details
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017098.1 | NP_058794.1 | 1 | 70 | ||
GenBank mRNA | L22788.1 | - | 1 | 70 | ||
S52878.1 | - | 1 | 49 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017098.1 | NP_058794.1 | 1 | 70 | ||
GenBank mRNA | L22788.1 | - | 1 | 70 | ||
S52878.1 | - | 1 | 49 |