siRNA Details
Gene Symbol: Fshb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007597.1 | NP_001007598.1 | 1 | 21 | ||
XM_006234672.2 | XP_006234734.1 | 2 | 1096 | |||
GenBank mRNA | BC168724.1 | - | 2 | 83 | 6889701 | |
M36804.1 | - | 1 | 34 |
