siRNA Details
Gene Symbol: Cenpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012955.1 | NP_037087.1 | 20 | 2082 | ||
XM_006257215.2 | XP_006257277.1 | 20 | 2220 | |||
XM_006257216.2 | XP_006257278.1 | 20 | 2381 | |||
XM_006257217.2 | XP_006257279.1 | 19 | 2095 | |||
XM_006257218.1 | XP_006257280.1 | 18 | 1747 | |||
XM_008773403.1 | XP_008771625.1 | 19 | 2436 | |||
GenBank mRNA | X90355.1 | - | 20 | 2082 |
