siRNA Details
Gene Symbol: Kcne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012973.1 | NP_037105.1 | 2 | 127 | ||
XM_006248034.2 | XP_006248096.1 | 2 | 140 | |||
XM_006248035.2 | XP_006248097.1 | 2 | 294 | |||
XM_008768566.1 | XP_008766788.1 | 2 | 511 | |||
GenBank mRNA | M22412.1 | - | 2 | 127 | ||
M36461.1 | - | 1 | 104 |
