siRNA Details
Gene Symbol: Lypla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013006.1 | NP_037138.1 | 8, 9 | 719 | ||
XM_008763522.1 | XP_008761744.1 | 7, 8 | 711 | |||
GenBank mRNA | BC085750.1 | - | 8, 9 | 657 | 7190638 | |
D63885.1 | - | 8, 9 | 719 | |||
U97146.1 | - | 8, 9 | 747 |
