siRNA Details
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013074.1 | NP_037206.1 | 2 | 444 | ||
XM_008766215.1 | XP_008764437.1 | 2 | 804 | |||
GenBank mRNA | AF041246.1 | - | 2 | 444 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013074.1 | NP_037206.1 | 2 | 444 | ||
XM_008766215.1 | XP_008764437.1 | 2 | 804 | |||
GenBank mRNA | AF041246.1 | - | 2 | 444 |
Set Membership: |
![]() |